Learn More
Sino Biological Anti-Factor XII Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 107065T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human Factor XII heavy chain, and purified by antigen affinity chromatography.
Catalog No. 50-240-8620