Learn More
Sino Biological Recombinant Human PAR2 Protein (VLP, Full-Length, His & FLAG Tag)
Catalog No. 502993484
Encompass_Preferred
A DNA sequence encoding the Human F2RL1-VLP (Full Length) (P55085) (Ile26-Tyr397) was expressed, with a Flag tag at the N-terminus, a polyhistidine tag and a strep tag at the C-terminus.
Catalog No. 50-299-3484 Supplier Sino Biological Supplier No. 13536H18H20
A DNA sequence encoding the Human F2RL1-VLP (Full Length) (P55085) (Ile26-Tyr397) was expressed, with a Flag tag at the N-terminus, a polyhistidine tag and a strep tag at the C-terminus.