Learn More
Sino Biological Anti-Tissue Factor Antibody (APC), Mouse Monoclonal
Supplier:  Sino Biological 13133MM05A100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human F3 / Tissue factor / CD142 (rh F3 / Tissue factor / CD142; Catalog#13133-H08H; P13726; Met 1-Glu 251) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-240-8020