Learn More
Sino Biological Anti-Tissue Factor Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 13133T52100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Coagulation Factor III / Tissue Factor / CD142 (rh Coagulation Factor III / Tissue Factor / CD142; Catalog#13133-H08H; P13726; Met1-Glu251). Coagulation Factor III / Tissue Factor / CD142 specific IgG was purified by Human Coagulation Factor III / Tissue Factor / CD142 affinity chromatography.
Catalog No. 50-241-1001