Learn More
Sino Biological Recombinant Human Tissue Factor Protein (hFc & AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 13133H41HB20
Encompass_Preferred
A DNA sequence encoding the Human F3 (P13726) (Met1-Glu251) was expressed with a c-terminal AVI tagged Fc region of human IgG1 at the C-terminus (Fc-AVI). The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-241-1270