Learn More
Sino Biological Human F9 ELISA Pair Set 15PP (Sino Biological)
Catalog No. 502370906
Encompass_Preferred
Each vial contains 240 ng of recombinant Human Coagulation Factor IX / FIX / F9. Reconstitute with 1 mL Dilution Buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in Dilution Buffer, and a high standard of 9000 pg/mL is recommended.
Catalog No. 50-237-0906 Supplier Sino Biological Supplier No. SEK1150315
Each vial contains 240 ng of recombinant Human Coagulation Factor IX / FIX / F9. Reconstitute with 1 mL Dilution Buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in Dilution Buffer, and a high standard of 9000 pg/mL is recommended.