Learn More
Sino Biological Human F9 MAb 100ul (Sino Biological)
Supplier:  Sino Biological 80715R104100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human Coagulation Factor IX / FIX / F9 (rh Coagulation Factor IX / FIX / F9; Catalog#11503-H08H; AAB59620.1; Met1-Thr461).
Catalog No. 50-237-4186