Learn More
Biotang Inc Human Folic acid,FA ELISA Kit (0.4ng/ml-20ng/ml), 96Test
Supplier:  Biotang Inc HU9161A
Encompass_Preferred
Human Folic acid,FA ELISA Kit (0.4ng/ml-20ng/ml), 96Test. Lead time of this kit is 1-2 weeks. Shipped by FedEx air. Storage: 2 - 8 C. Shelf Time: 6 months. Kit Experimental Time:< 2 hs. TSZELISA brand.
Catalog No. 50-153-079