Learn More
Raybiotech Inc Human FABP5 (E-FABP) ELISA Kit
Supplier:  Raybiotech Inc ELHFABP51
Encompass_Preferred
For serum, plasma, and cell culture supernatant samples. Sensitivity: 12.5 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects human FABP-5. Other species not determined.
Catalog No. 50-194-9238