Learn More
Raybiotech Inc Human FABP6 ELISA Kit
Supplier:  Raybiotech Inc ELHFABP61
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 1.64 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human FABP6.
Catalog No. 50-194-9239