Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human FABP6 ELISA Kit

Catalog No. EH179RBX5
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH179RBX5 5 x 96 Tests
EH179RB 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EH179RBX5 Supplier Invitrogen™ Supplier No. EH179RBX5
Get Up to $3,000 in Extra Life Sciences Consumables with Purchase
View all promotions
Add to Cart
Add to Cart

ELISA

Human FABP6 quantitates human FABP6 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human FABP6.

This gene encodes the ileal fatty acid binding protein. Fatty acid binding proteins are a family of small, highly conserved, cytoplasmic proteins that bind long-chain fatty acids and other hydrophobic ligands. FABP6 and FABP1 (the liver fatty acid binding protein) are also able to bind bile acids. It is thought that FABPs roles include fatty acid uptake, transport, and metabolism. Transcript variants generated by alternate transcription promoters and/or alternate splicing have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P51161
Assay Range 1.64-400 ng/mL
Assay Sensitivity 1.64 ng/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias FABP6; fatty acid binding protein 6; fatty acid binding protein 6, ileal; fatty acid binding protein 6, ileal (gastrotropin); fatty acid-binding protein 6; Gastrotropin; GT; I-15P; I-BABP; I-BALB; I-BAP; ILBP; ILBP3; ileal bile acid binding protein; ileal bile acid-binding protein; ileal lipid-binding protein; Illbp; illeal lipid-binding protein; intestinal 15 kDa protein; Intestinal bile acid-binding proteinFABP6; fatty acid binding protein 6; fatty acid binding protein 6, ileal; fatty acid binding protein 6, ileal (gastrotropin); fatty acid-binding protein 6; Gastrotropin; GT; I-15P; I-BABP; I-BALB; I-BAP; ILBP; ILBP3; ileal bile acid binding protein; ileal bile acid-binding protein; ileal lipid-binding protein; Illbp; illeal lipid-binding protein; intestinal 15 kDa protein; Intestinal bile acid-binding protein;
Gene ID (Entrez) 2172
Gene Symbol FABP6
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target FABP6
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.