Learn More
BIOHIPPO Human FABP7 Microsample
SDP
Catalog No. 5000314635
Encompass
Human FABP7 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of FABP7 in Serum plasma and other biological fluids It provides a detection range of 0 47-30 ng/mL with a sensitivity of 0 19 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human FABP7 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human FABP7 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human FABP7 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14635 Supplier BIOHIPPO Supplier No. BHE15212180
Human FABP7 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of FABP7 in Serum plasma and other biological fluids It provides a detection range of 0 47-30 ng/mL with a sensitivity of 0 19 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human FABP7 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human FABP7 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human FABP7 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color