Learn More
Raybiotech Inc Human FABP8/M-FABP ELISA Kit
Supplier:  Raybiotech Inc ELHFABP81
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 4.1 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human FABP8.
Catalog No. 50-194-9240