Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human FABP8 (M-FABP) Recombinant Protein

Catalog No. PIRP75557
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIRP75557 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIRP75557 Supplier Invitrogen™ Supplier No. RP75557
Only null left
Add to Cart
Add to Cart

Recombinant Protein

This product contains purified recombinant human myelin FABP protein with an N-terminal hexahistidine tag. This product may be used as a positive control in many immunological assays.

The protein encoded by this gene localizes to myelin sheaths of the peripheral nervous system. The encoded protein can bind both the membrane layers of the sheaths and monomeric lipids, and is thought to provide stability to the sheath. A defect in this gene was shown to be a cause of dominant demyelinating CMT neuropathy.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P02689
Concentration 0.22 mg/mL
For Use With (Application) Control, Western Blot
Formulation 20 mM tris HCl with 50% glycerol and no preservative; pH 8
Gene ID (Entrez) 5375
Name Human FABP8 (M-FABP)
Quantity 10 μg
Source E. coli
Immunogen N/A
Regulatory Status RUO
Gene Alias FABP8; M FABP8; M-FABP; MP2; myelin P2 protein; P2; Peripheral myelin protein 2; PMP2
Common Name FABP8
Gene Symbol FABP4
Conjugate Unconjugated
Species E. coli
Recombinant Recombinant
Protein Tag His-tag
Sequence Amino acids Met1-Val132 containing an N-terminal His-tag
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Protein Subtype Recombinant
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.