Learn More
Enzyme Research Laboratories Inc HUMAN FACTOR 10; 0.8MG
Catalog No. NC9823146
Encompass
Human Factor X,100 U (0.8 mg),Buffer Composition = 20 mM Tris-HCl,0.1 M NaCl,1 mM Benzamidine,pH 7.4,Extinction Coefficient (1%) = 11.6,One Unit = 8 ug,Molecular Weight = 58,800 daltons,Prepared from fresh frozen human plasma
Catalog No. NC9823146 Supplier Enzyme Research Laboratories Inc Supplier No. HFX 1010
Human Factor X,100 U (0.8 mg),Buffer Composition = 20 mM Tris-HCl,0.1 M NaCl,1 mM Benzamidine,pH 7.4,Extinction Coefficient (1%) = 11.6,One Unit = 8 ug,Molecular Weight = 58,800 daltons,Prepared from fresh frozen human plasma