Learn More
Enzyme Research Laboratories Inc HUMAN FACTOR ALPHA XIIA 0.5MG
Catalog No. NC9679522
Encompass
Human Factor alpha -XIIa is a serine protease responsible for the activation of Factor XI to XIa in the contact activation system. The protein purity is determined by SDS-PAGE and activity is determined via clotting assay.
This item is not returnable. View return policy
Catalog No. NC9679522 Supplier Enzyme Research Laboratories Inc Supplier No. HFXIIA 1212A
Human Factor alpha -XIIa is a serine protease responsible for the activation of Factor XI to XIa in the contact activation system. The protein purity is determined by SDS-PAGE and activity is determined via clotting assay.