Learn More
Enzyme Research Laboratories Inc HUMAN FACTOR ALPHAXIIA 1MG
Catalog No. NC0360100
Encompass
Human Factor alpha -XIIa is a serine protease responsible for the activation of Factor XI to XIa in the contact activation system. The protein purity is determined by SDS-PAGE and activity is determined via clotting assay.
Catalog No. NC0360100 Supplier Enzyme Research Laboratories Inc Supplier No. HFXIIA 1212A1MG
Human Factor alpha -XIIa is a serine protease responsible for the activation of Factor XI to XIa in the contact activation system. The protein purity is determined by SDS-PAGE and activity is determined via clotting assay.