Learn More
Abcam Human Factor IX ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB1088311X96TESTS
Encompass_Preferred
Human Factor IX ELISA Kit ab108831 is used to quantify Human Factor IX with a sensitivity of 20.6 pg/ml. Sandwich ELISA Kit
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1983576