Learn More
Biomatik Corporation Human Factor Related Apoptosis (FAS) ELISA Kit
Supplier:  Biomatik Corporation EKU04010
Encompass_Preferred
Human Factor Related Apoptosis (FAS) ELISA Kit for detecting FAS in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 39-2,500pg/ml. Sensitivity: 15pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P25445. Gene ID: 355. Target Synonyms: CD95; ALPS1A; ALPS1-A; APO1; APT1; FAS1; FASTM; TNFRSF6; Fas Receptor; TNF Receptor Superfamily Member 6; Tumor Necrosis Factor Receptor Superfamily Member 6. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4960