Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Factor V Native Protein

Catalog No. PIRP43126 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIRP43126 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIRP43126 Supplier Invitrogen™ Supplier No. RP43126
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Native Protein

RP-43126 contains native human Factor V protein. RP-43126 may be used as a positive control in many immunological assays. The product is of human blood/plasma origin. Although the starting material was tested prior to initiation of the manufacturing process, and was found negative or nonreactive for anti-HIV-1/2, HIV-1 antigen(s), HBsAg, STS, anti-HCV, anti-HBcore and anti-HTLV I & II, extreme caution should be used when handling this material as there is a margin of error in all tests. This product is to be used for IN VITRO research purposes only, and is not intended for clinical or diagnostic use!.

Coagulation factor V (or Proaccelerin) is an essential factor of the blood coagulation cascade. This factor circulates in plasma, and is converted to the active form by the release of the activation peptide by thrombin during coagulation. This generates a heavy chain and a light chain which are held together by calcium ions. The active factor V is a cofactor that participates with activated coagulation factor X to activate prothrombin to thrombin. Defects in the Factor V gene can result various diseases including Factor V deficiency, thrombophilia due to activated protein C resistance, Budd-Chiari syndrome, Ischemic stroke, and recurrent pregnancy loss.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P12259
Concentration 5.2 mg/mL
For Use With (Application) Neutralization
Formulation 50% water with 50% glycerol and no preservative
Gene ID (Entrez) 2153
Molecular Weight (g/mol) 330 kDa
Name Human Factor V
Purification Method SDS-PAGE
Quantity 50 μg
Source Human
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias Ac2-120; Activated protein C cofactor; AI173222; Cf5; Cf-5; coagulation factor V; coagulation factor V (proaccelerin, labile factor); Coagulation factor V heavy chain; coagulation factor V jinjiang A2 domain; Coagulation factor V light chain; F5; Factor 5; factor V; factor V Leiden; Factor5; FVL; PCCF; Proaccelerin, labile factor; RPRGL1; THPH2
Common Name Factor V
Gene Symbol F5
Product Type Protein
Conjugate Unconjugated
Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Form Liquid
Protein Subtype Native
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.