Learn More
Haematologic Technologies Inc HUMAN FACTOR XI 50UG
Supplier:  Haematologic Technologies Inc HCXI0150
Encompass_Preferred
Coagulation Protein, Human Factor XI, 50 micrograms, Purifed from plasma, MW=160,000, Structure: homodimer consisting of two apparently identical subunits (Mr~80,000) held together by disulfide bonds, RUO, 50% (v/v) glycerol/water, Store at -20 C.
Catalog No. 50-883-391