Learn More
Haematologic Technologies Inc HUMAN FACTOR XIA 50UG
Supplier:  Haematologic Technologies Inc HCXIA0160
Encompass_Preferred
Coagulation protein, Human factor XIa, 50 micrograms, Enzyme, Mode of action: A serine protease that converts factor IX to factor IXa, Research use only, 50% (v/v) glycerol/water, store at -20 Celsius
Catalog No. 50-883-422