Learn More
American Research Products Inc Human FAD linked sulfhydryl oX
Catalog No. 5000327042
Encompass
Human FAD-linked sulfhydryl oXidase ALR (GFER) ELISA kit from CUSABIO Detection Range 1 56 pg/ml-100 pg/ml Sensitivity 0 39 pg/ml Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27042 Supplier American Research Products Inc Supplier No. CSBEL009370HU
Human FAD-linked sulfhydryl oXidase ALR (GFER) ELISA kit from CUSABIO Detection Range 1 56 pg/ml-100 pg/ml Sensitivity 0 39 pg/ml Sample serum plasma tissue homogenates Method Sandwich