Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human FAIM Synthetic Peptide

Catalog No. PEP0432
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP0432 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP0432 Supplier Invitrogen™ Supplier No. PEP0432
Only null left
Add to Cart
Add to Cart

Synthetic Peptide

FAIM Synthetic Peptide, PEP-0432, from Invitrogen.

Plays a role as an inducible effector molecule that mediates Fas resistance produced by surface Ig engagement in B cells.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 55179
Species Human
Purification Method purified
Accession Number Q9NVQ4
Common Name Human FAIM
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Symbol FAIM
Lyophilized No
Preservative 0.02% sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.