Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human FAIM3 Synthetic Peptide

Catalog No. PEP0403 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP0403 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP0403 Supplier Invitrogen™ Supplier No. PEP0403
Add to Cart
Add to Cart

Synthetic Peptide

This peptide corresponds to 13 amino acids near the carboxy terminus of human Toso. PEP-0403 can be used as a blocking peptide with polyclonal antibody PA5-20285.

Apoptosis is an important process by which normal tissue homeostasis and function are maintained. One of the major signals that regulate this process is mediated by the activation of the Fas receptor by its ligand. This leads to the formation of a Fas-associated death domain (FADD)- containing death-inducing signaling complex and the activation of caspase-8, which in turn activates downstream effector caspases, such as caspase-3 and -7. Recent experiments have shown that overexpression of Toso, a novel regulator of Fas-induced apoptosis in lymphoid cells, in Jurkat cells as well as transgenic mice render these cells resistant to Fas-induced apoptosis but not to TRAIL-induced apoptosis. Furthermore, Toso was found to associate with FADD, suggesting that Toso functions by disrupting the formation of the death-inducing signaling complex.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 9214
Species Human
Purification Method purified
Accession Number O60667
Common Name FAIM3
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Alias 1810037B05Rik; Faim3; Fas apoptotic inhibitory molecule 3; Fc fragment of IgM receptor; Fc mu receptor; FCMR; FcmuR; IgM Fc fragment receptor; IgM Fc receptor; immunoglobulin mu Fc receptor; regulator of Fas-induced apoptosis Toso; TOSO
Gene Symbol Fcmr
Lyophilized No
Preservative Sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.