Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human FAM193B Synthetic Peptide

Catalog No. PEP1482
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP1482 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP1482 Supplier Invitrogen™ Supplier No. PEP1482
  • $213.00
    $174.60 / Each of 1

    Save $38.40

    18% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

Synthetic Peptide

FAM193B Synthetic Peptide, PEP-1482, from Invitrogen.

FAM193B, also known as IRIZIO, was initially identified as a protein that is upregulated in alveolar rhabdomyosarcoma (ARMS), a type of fast-growing tumor characterized by chromosomal translocations fusing the PAX3 or PAX7 gene with that of FOXO1. It has been suggested that, in addition to the PAX3-FOXO1 gene fusion, disruption of the Rb and p53 pathways is required for full ARMS tumorigenesis. In PAX3-FOXO1-expressing primary mouse fibroblasts that possessed a defective p53 pathway, FAM193B expression enabled the tumorigenic transformation, suggesting that FAM193B may contribute to rhabdomyosarcomagenesis in humans.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 54540
Species Human
Purification Method purified
Accession Number Q96PV7
Common Name Human FAM193B
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Symbol FAM193B
Lyophilized No
Preservative 0.02% sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.