Learn More
Biomatik Corporation Human Family With Sequence Similarity 19, Member A4 (FAM19A4) ELISA Kit
Supplier:  Biomatik Corporation EKU12496
Encompass_Preferred
Human Family With Sequence Similarity 19, Member A4 (FAM19A4) ELISA Kit for detecting FAM19A4 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q96LR4. Gene ID: 151647. Target Synonyms: TAFA-4; Chemokine-like protein TAFA-4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5936