Learn More
ACROBiosystems PE-Labeled Human FAP, His Tag (FAP-HP245) is produced via site-specific conjugation of PE to Human FAP, His Tag under optimal conditions with a proprietary technology.It contains AA Leu 26 - Asp 760 (Accession # Q12884-1).
Supplier:  ACROBiosystems FAPHP245200TESTS
Encompass_Preferred
FAP (also known as seprase) is a Type II transmembrane serine protease. Both plasma membrane and soluble forms exhibit post-proline cleaving endopeptidase activity, with a marked preference for Ala/Ser-Gly-Pro-Ser/Asn/Ala consensus sequences. Degrade also gelatin, heat-denatured type I collagen. Also has dipeptidyl peptidase activity, with a preference for Ala-Pro, Ile-Pro, Gly-Pro, Arg-Pro and Pro-Pro. The plasma membrane form, in association with either DPP4, PLAUR or integrins, is involved in the pericellular proteolysis of the extracellular matrix (ECM), and hence promotes cell adhesion, migration and invasion through the ECM. Promotes glioma cell invasion through the brain parenchyma by degrading the proteoglycan brevican. Acts as a tumor suppressor in melanocytic cells through regulation of cell proliferation and survival in a serine protease activity-independent manner.
Catalog No. 50-223-1278