Learn More
ACROBiosystems Human FAP 2mg
Catalog No. 502534451
Encompass_Preferred
The biotinylated FAP protein was conjugated to streptavidin magnetic beads. This pre-coupled magnetic bead product can capture the anti-FAP antibody from various assay systems. The beads are in uniform size narrow size distribution with large surface area and unique surface coating which can help you get the best performance and highly reproducible results. This FAP coupled magnetic beads will bring great convenience with minimum non-specific binding and developed protocols. This ready-to-use product could greatly save your time and hassle.
Catalog No. 50-253-4451 Supplier ACROBiosystems Supplier No. MBSK0772MG
The biotinylated FAP protein was conjugated to streptavidin magnetic beads. This pre-coupled magnetic bead product can capture the anti-FAP antibody from various assay systems. The beads are in uniform size narrow size distribution with large surface area and unique surface coating which can help you get the best performance and highly reproducible results. This FAP coupled magnetic beads will bring great convenience with minimum non-specific binding and developed protocols. This ready-to-use product could greatly save your time and hassle.