Learn More
Raybiotech Inc HUMAN FAP ELISA
Supplier:  Raybiotech Inc ELHFAP1
Encompass_Preferred
RayBio Human FAP ELISA Kit for cell culture supernatants, plasma, and serum samples. Pre-coated 96-well microplate kit with additional reagents. Detection Range: 12 pg/ml - 4000 pg/ml
Catalog No. 50-104-3758