Learn More
Sino Biological Human FAP PAb 100ul (Sino Biological)
Supplier:  Sino Biological 10464T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human FAP (rh FAP; Catalog#10464-H07H; Q12884-1; Leu26-Asp760). FAP specific IgG was purified by Human FAP affinity chromatography.
Catalog No. 50-237-2017