Learn More
Sino Biological Human FAP Protein 50ug (Sino Biological)
Supplier:  Sino Biological 10464H07B50
Encompass_Preferred
A DNA sequence encoding the human FAP (Q12884-1) (Arg27-Asp760) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-237-3028