Learn More
Sino Biological Recombinant Human FAP Protein (ECD, His & AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 10464H56HB20
Encompass_Preferred
A DNA sequence encoding the Human FAP (Q12884-1) (Leu26-Asp760) was expressed with a N-terminal polyhistidine tag followed by an AVI tag.The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-240-8477