Learn More
Biomatik Corporation Human Far Upstream Element Binding Protein 1 (FUBP1) ELISA Kit
Supplier:  Biomatik Corporation EKU11781
Encompass_Preferred
Human Far Upstream Element Binding Protein 1 (FUBP1) ELISA Kit for detecting FUBP1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78-5,000pg/ml. Sensitivity: 33pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q96AE4. Gene ID: 8880. Target Synonyms: FBP; FUBP; hDH V; DNA helicase V. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7216