Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human FARSA Full-length ORF (NP_004452.1, 1 a.a. - 508 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89944080 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-944-080 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-944-080 Supplier Abnova™ Supplier No. H00002193P01
Only null left
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

Aminoacyl-tRNA synthetases are a class of enzymes that charge tRNAs with their cognate amino acids. This gene encodes a product which is similar to the catalytic subunit of prokaryotic and Saccharomyces cerevisiae phenylalanyl-tRNA synthetases (PheRS). This gene product has been shown to be expressed in a tumor-selective and cell cycle stage- and differentiation-dependent manner, the first member of the tRNA synthetase gene family shown to exhibit this type of regulated expression [provided by RefSeq]

Sequence: MADGQVAELLLRRLEASDGGLDSAELAAELGMEHQAVVGAVKSLQALGEVIEAELRSTKHWELTAEGEEIAREGSHEARVFRSIPPEGLAQSELMRLPSGKVGFSKAMSNKWIRVDKSAADGPRVFRVVDSMEDEVQRRLQLVRGGQAEKLGEKERSELRKRKLLAEVTLKTYWVSKGSAFSTSISKQETELSPEMISSGSWRDRPFKPYNFLAHGVLPDSGHLHPLLKVRSQFRQIFLEMGFTEMPTDNFIESSFWNFDALFQPQQHPARDQHDTFFLRDPAEALQLPMDYVQRVKRTHSQGGYGSQGYKYNWKLDEARKNLLRTHTTSASARALYRLAQKKPFTPVKYFSIDRVFRNETLDATHLAEFHQIEGVVADHGLTLGHLMGVLREFFTKLGITQLRFKPAYNPYTEPSMEVFSYHQGLKKWVEVGNSGVFRPEMLLPMGLPENVSVIAWGLSLERPTMIKYGINNIRELVGHKVNLQMVYDSPLCRLDAEPRPPPTQEAA

Specifications

Accession Number NP_004452.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 2193
Molecular Weight (g/mol) 84kDa
Name FARSA (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 μg
Immunogen MADGQVAELLLRRLEASDGGLDSAELAAELGMEHQAVVGAVKSLQALGEVIEAELRSTKHWELTAEGEEIAREGSHEARVFRSIPPEGLAQSELMRLPSGKVGFSKAMSNKWIRVDKSAADGPRVFRVVDSMEDEVQRRLQLVRGGQAEKLGEKERSELRKRKLLAEVTLKTYWVSKGSAFSTSISKQETELSPEMISSGSWRDRPFKPYNFLAHGVLPDSGHLHPLLKVRSQFRQIFLEMGFTEMPTDNFIESSFWNFDALFQPQQHPARDQHDTFFLRDPAEALQLPMDYVQRVKRTHSQGGYGSQGYKYNWKLDEARKNLLRTHTTSASARALYRLAQKKPFTPVKYFSIDRVFRNETLDATHLAEFHQIEGVVADHGLTLGHLMGVLREFFTKLGITQLRFKPAYNPYTEPSMEVFSYHQGLKKWVEVGNSGVFRPEMLLPMGLPENVSVIAWGLSLERPTMIKYGINNIRELVGHKVNLQMVYDSPLCRLDAEPRPPPTQEAA
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias CML33/FARSL/FARSLA/FRSA/PheHA
Common Name FARSA
Gene Symbol FARSA
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.