Learn More
Biomatik Corporation Human Fas Associating Death Domain Containing Protein (FADD) ELISA Kit
Supplier:  Biomatik Corporation EKU10104
Encompass_Preferred
Human Fas Associating Death Domain Containing Protein (FADD) ELISA Kit for detecting FADD in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.065ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q13158. Gene ID: 8772. Target Synonyms: GIG3; MORT1; FADD Death Effector Domain; Fas-Associated Protein With Death Domain; Mediator Of Receptor-Induced Toxicity; Growth-Inhibiting Gene 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5537