Learn More
Sino Biological Human Fas/CD95/TNFRSF6 Matched ELISA Antibody Pair Set 5 Plates
Catalog No. 5000786053
Encompass_Preferred
The Human CD95 / APO-1 / TNFRSF6 / FAS ELISA Pair Set is for the quantitative determination of Human CD95 / APO-1 / TNFRSF6 / FAS.This ELISA Pair Set contains the basic components required for the development of sandwich ELISAs.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for CD95 / APO-1 / TNFRSF6 / FAS coated on a 96-well plate. Standards and samples are added to the wells, and any CD95 / APO-1 / TNFRSF6 / FAS present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated mouse anti-CD95 / APO-1 / TNFRSF6 / FAS monoclonal antibody is then added, producing an antibody-antigen-antibody “sandwich”. The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of CD95 / APO-1 / TNFRSF6 / FAS present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the mic
Catalog No. 50-007-86053 Supplier Sino Biological Supplier No. SEK102175
The Human CD95 / APO-1 / TNFRSF6 / FAS ELISA Pair Set is for the quantitative determination of Human CD95 / APO-1 / TNFRSF6 / FAS.This ELISA Pair Set contains the basic components required for the development of sandwich ELISAs.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for CD95 / APO-1 / TNFRSF6 / FAS coated on a 96-well plate. Standards and samples are added to the wells, and any CD95 / APO-1 / TNFRSF6 / FAS present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated mouse anti-CD95 / APO-1 / TNFRSF6 / FAS monoclonal antibody is then added, producing an antibody-antigen-antibody “sandwich”. The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of CD95 / APO-1 / TNFRSF6 / FAS present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the mic