Learn More
Raybiotech Inc Human Fas (TNFRSF6/Apo-1) ELISA Kit
Supplier:  Raybiotech Inc ELHFASCL1
Encompass_Preferred
For cell and tissue lysate samples. Sensitivity: 5 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: The antibody pair provided in this kit recognizes human Fas.
Catalog No. 50-194-9243