Learn More
Biomatik Corporation Human Fasciculation And Elongation Protein Zeta 1 (FEz1) ELISA Kit
Supplier:  Biomatik Corporation EKU12523
Encompass_Preferred
Human Fasciculation And Elongation Protein Zeta 1 (FEz1) ELISA Kit for detecting FEz1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q99689. Gene ID: 9638. Target Synonyms: FE-Z1; Zygin I. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6691