Learn More
Sino Biological Anti-Fas Ligand/TNFSF6 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10244T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Fas Ligand/FASLG (rh Fas Ligand/FASLG; Catalog#10244-H07Y; NP_000630.1; Pro134-Leu281). Fas Ligand/FASLG specific IgG was purified by Human Fas Ligand/FASLG affinity chromatography.
Catalog No. 50-241-1440