Learn More
Sino Biological Human FASN Protein 500ug (Sino Biological)
Supplier:  Sino Biological 30113H07E500
Encompass_Preferred
A DNA sequence encoding the Human FASN (P49327) (Leu2200-Gly2511) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-237-1796