Learn More
BIOHIPPO Human FATP5 Microsample
SDP
Catalog No. 5000314546
Encompass
Human FATP5 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of FATP5 in Tissue homogenates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 119 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human FATP5 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human FATP5 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human FATP5 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14546 Supplier BIOHIPPO Supplier No. BHE15211780
Human FATP5 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of FATP5 in Tissue homogenates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 119 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human FATP5 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human FATP5 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human FATP5 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color