Learn More
Biomatik Corporation Human Fatty Acid Binding Protein 5 (FABP5) CLIA Kit
Supplier:  Biomatik Corporation EKU09779
Encompass_Preferred
Human Fatty Acid Binding Protein 5 (FABP5) CLIA Kit for detecting FABP5 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 54.9-40,000pg/ml. Sensitivity: 19.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q01469. Gene ID: 2171. Target Synonyms: E-FABP; EFABP; PA-FABP; PAFABP; Fatty Acid Binding Protein 5, Epidermal; Psoriasis-Associated; Epidermal-type fatty acid-binding protein; Psoriasis-associated fatty acid-binding protein homolog. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4788