Learn More
Biomatik Corporation Human Fatty Acid Desaturase 3 (FADS3) ELISA Kit
Supplier:  Biomatik Corporation EKU04051
Encompass_Preferred
Human Fatty Acid Desaturase 3 (FADS3) ELISA Kit for detecting FADS3 in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.118ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9Y5Q0. Gene ID: 3995. Target Synonyms: CYB5RP; Delta-9-Desaturase; Cytochrome b5-related protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7617