Learn More
Abcam Human Fatty Acid Oxidation In-Cell ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB1181821X96TESTS
Encompass_Preferred
Human Fatty Acid Oxidation In-Cell ELISA Kit is Cell-based (quantitative) ELISA for the measurement of Human Fatty Acid Oxidation production in in Suspension cells, Adherent cells samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2107176