Learn More
Biomatik Corporation Human F-Box Protein 7 (FBXO7) ELISA Kit
Supplier:  Biomatik Corporation EKU12044
Encompass_Preferred
Human F-Box Protein 7 (FBXO7) ELISA Kit for detecting FBXO7 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.061ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9Y3I1. Gene ID: 25793. Target Synonyms: FBX; FBX07; FBX7. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5720