Learn More
ACROBiosystems Biotinylated Human Fc epsilon RI alpha Protein, His,Avitag™ (MALS & BLI verified)
Catalog No. 502719646
Encompass_Preferred
High affinity immunoglobulin epsilon receptor subunit alpha (FCER1A) is also known as Fc-epsilon RI-alpha (FcERI), IgE Fc receptor subunit alpha, FCE1A. FCER1A contains two Ig-like (immunoglobulin-like) domains. FCER1A binds to the Fc region of immunoglobulins epsilon and is a high affinity receptor. FCER1A is responsible for initiating the allergic response, which binding of allergen to receptor-bound IgE leads to cell activation and the release of mediators (such as histamine) responsible for the manifestations of allergy. The same receptor also induces the secretion of important lymphokines. FCER1A plays a central role in allergic disease, coupling allergen and mast cell to initiate the inflammatory and immediate hypersensitivity responses that are characteristic of disorders such as hay fever and asthma.
Catalog No. 50-271-9646 Supplier ACROBiosystems Supplier No. FCAH82E325UG
 Restricted Item
Add to Cart
Add to Cart
High affinity immunoglobulin epsilon receptor subunit alpha (FCER1A) is also known as Fc-epsilon RI-alpha (FcERI), IgE Fc receptor subunit alpha, FCE1A. FCER1A contains two Ig-like (immunoglobulin-like) domains. FCER1A binds to the Fc region of immunoglobulins epsilon and is a high affinity receptor. FCER1A is responsible for initiating the allergic response, which binding of allergen to receptor-bound IgE leads to cell activation and the release of mediators (such as histamine) responsible for the manifestations of allergy. The same receptor also induces the secretion of important lymphokines. FCER1A plays a central role in allergic disease, coupling allergen and mast cell to initiate the inflammatory and immediate hypersensitivity responses that are characteristic of disorders such as hay fever and asthma.