Learn More
ACROBiosystems ACROBiosystems Human Fc epsilon RI alpha Protein, His Tag
Catalog No. 502106636
Encompass_Preferred
Recombinant Protein;1MG;Human Fc epsilon RI alpha, His Tag (FCA-H5228) is expressed from human 293 cells (HEK293). It contains AA Val 26 - Gln 205 (Accession # P12319-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 22.9 kDa. The protein migrates as 37-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Immunoglobulin E, Human Plasma at 5 μg/mL (100 μL/well) can bind Human Fc epsilon RI alpha, His Tag (Cat. No. FCA-H5228) with a linear range of 0.4-3 ng/mL (QC tested).
Catalog No. 50-210-6636 Supplier ACROBiosystems Supplier No. FCAH52281MG
Recombinant Protein;1MG;Human Fc epsilon RI alpha, His Tag (FCA-H5228) is expressed from human 293 cells (HEK293). It contains AA Val 26 - Gln 205 (Accession # P12319-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 22.9 kDa. The protein migrates as 37-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Immunoglobulin E, Human Plasma at 5 μg/mL (100 μL/well) can bind Human Fc epsilon RI alpha, His Tag (Cat. No. FCA-H5228) with a linear range of 0.4-3 ng/mL (QC tested).