Learn More
ACROBiosystems HUMAN FC GAMMA RI CD64 PROTEIN
Catalog No. NC1824662
Encompass_Preferred
Human Fc gamma RI / CD64 Protein, His Tag (SPR & BLI verified). Lyophilized from 0.22 m filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
Catalog No. NC1824662 Supplier ACROBiosystems Supplier No. FCAH52H150UG
Human Fc gamma RI / CD64 Protein, His Tag (SPR & BLI verified). Lyophilized from 0.22 m filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.