Learn More
ACROBiosystems Human Fc gamma RIIB / CD32b Protein, SUMO,His Tag
Supplier:  ACROBiosystems CDBH5298100UG
Encompass_Preferred
Recombinant Protein;100UG;Human CD32b Protein, SUMO,His Tag (CDB-H5298) is expressed from human 293 cells (HEK293). It contains AA Ala 46 - Pro 217 (Accession # P31994-1).;This protein carries a SUMO tag at the N-terminus and a polyhistidine tag at the C-terminus. The protein has a calculated MW of 32.0 kDa. The protein migrates as 40-45 kDa under reducing (R) condition (SDS-PAGE) due to Glycosylation.
Catalog No. 50-205-4988